Assay Details
| Gene Symbol: Akt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110208.1 | NP_001103678.1 | 3 - 4 | 580 | 75 | |
| NM_007434.3 | NP_031460.1 | 3 - 4 | 338 | 75 | ||
| GenBank mRNA | AK144217.1 | - | 3 - 4 | 397 | 75 | |
| AK158792.1 | - | 3 - 4 | 581 | 75 | ||
| AK191996.1 | - | 1 - 2 | 133 | 75 | ||
| AK197364.1 | - | 1 - 2 | 133 | 75 | ||
| BC026151.1 | - | 3 - 4 | 361 | 4187425 | 75 | |
| BC028759.1 | - | 2 - 3 | 254 | 75 | ||
| BC040377.1 | - | 3 - 4 | 330 | 4500854 | 75 | |
| U22445.1 | - | 3 - 4 | 391 | 75 | ||
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | AK050325.1 | - | 625 | Chr. - - [-] on Build | 75 | ||
More Information
Set Membership: |
|

Akt2
Genomic Map