Assay Details
| Gene Symbol: Capza1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009797.2 | NP_033927.2 | 1 - 2 | 164 | 78 | |
| GenBank mRNA | AK146296.1 | - | 1 - 2 | 50 | 78 | |
| AK147049.1 | - | 1 - 2 | 53 | 78 | ||
| AK149602.1 | - | 1 - 2 | 50 | 78 | ||
| AK152364.1 | - | 1 - 2 | 49 | 78 | ||
| AK163418.1 | - | 1 - 2 | 50 | 78 | ||
| AK165655.1 | - | 1 - 2 | 52 | 78 | ||
| AK166484.1 | - | 1 - 2 | 48 | 78 | ||
| AK166964.1 | - | 1 - 2 | 53 | 78 | ||
| AK167193.1 | - | 1 - 2 | 51 | 78 | ||
| AK168099.1 | - | 1 - 2 | 50 | 78 | ||
| AK168916.1 | - | 1 - 2 | 52 | 78 | ||
| AK169806.1 | - | 1 - 2 | 50 | 78 | ||
| BC016232.1 | - | 1 - 2 | 53 | 4489604 | 78 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK146296.1, AK147049.1, AK149602.1, AK152364.1, AK163418.1, AK166484.1, AK166964.1, AK167193.1, AK168916.1, AK169806.1, BC016232.1, NM_009797.2 transcripts.
Set Membership: |
|

Capza1
Genomic Map