Assay Details
| Gene Symbol: Capza1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009797.2 | NP_033927.2 | 2 - 3 | 228 | 95 | |
| GenBank mRNA | AK146296.1 | - | 2 - 3 | 114 | 95 | |
| AK147049.1 | - | 2 - 3 | 117 | 95 | ||
| AK149602.1 | - | 2 - 3 | 114 | 95 | ||
| AK151253.1 | - | 2 - 3 | 125 | 95 | ||
| AK152364.1 | - | 2 - 3 | 113 | 95 | ||
| AK163418.1 | - | 2 - 3 | 114 | 95 | ||
| AK165655.1 | - | 2 - 3 | 116 | 95 | ||
| AK166484.1 | - | 2 - 3 | 112 | 95 | ||
| AK166964.1 | - | 2 - 3 | 117 | 95 | ||
| AK167193.1 | - | 2 - 3 | 115 | 95 | ||
| AK168099.1 | - | 2 - 3 | 114 | 95 | ||
| AK168916.1 | - | 2 - 3 | 116 | 95 | ||
| AK169806.1 | - | 2 - 3 | 114 | 95 | ||
| BC016232.1 | - | 2 - 3 | 117 | 4489604 | 95 | |
| BC085506.1 | - | 2 - 3 | 103 | 6817308 | 95 | |
| U16740.1 | - | 2 - 3 | 97 | 95 | ||
More Information
Set Membership: |
|

Capza1
Genomic Map