Assay Details
| Gene Symbol: Capza1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009797.2 | NP_033927.2 | 5 - 6 | 549 | 107 | |
| GenBank mRNA | AK146296.1 | - | 5 - 6 | 435 | 107 | |
| AK147049.1 | - | 5 - 6 | 438 | 107 | ||
| AK149602.1 | - | 5 - 6 | 435 | 107 | ||
| AK151253.1 | - | 5 - 6 | 446 | 107 | ||
| AK152364.1 | - | 5 - 6 | 434 | 107 | ||
| AK161370.1 | - | 5 - 6 | 551 | 107 | ||
| AK163418.1 | - | 5 - 6 | 435 | 107 | ||
| AK165655.1 | - | 5 - 6 | 437 | 107 | ||
| AK166484.1 | - | 5 - 6 | 433 | 107 | ||
| AK166964.1 | - | 5 - 6 | 438 | 107 | ||
| AK167193.1 | - | 5 - 6 | 436 | 107 | ||
| AK168099.1 | - | 5 - 6 | 435 | 107 | ||
| AK168916.1 | - | 5 - 6 | 437 | 107 | ||
| AK169806.1 | - | 5 - 6 | 435 | 107 | ||
| BC016232.1 | - | 5 - 6 | 438 | 4489604 | 107 | |
| BC085506.1 | - | 5 - 6 | 424 | 6817308 | 107 | |
| U16740.1 | - | 5 - 6 | 418 | 107 | ||
More Information
Set Membership: |
|

Capza1
Genomic Map