Assay Details
| Gene Symbol: 5430406J06Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK017280.1 | - | 1 | 122 | 99 | |
| Gene Symbol: Ube2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK135056.1 | - | 1 | 3375 | 99 | |
More Information
Set Membership: |
|

5430406J06Rik
Genomic Map