Assay Details
| Gene Symbol: Trappc12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK045717.1 | - | 1422 | 70 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trappc12| Gene Symbol: Trappc12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK045717.1 | - | 1422 | 70 | ||