Assay Details
| Gene Symbol: Synj2bp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025292.6 | NP_079568.1 | 4 | 3036 | 96 | |
| GenBank mRNA | AK030379.1 | - | 3 | 2892 | 96 | |
| AK033514.1 | - | 4 | 3036 | 96 | ||
More Information
Set Membership: |
|

Synj2bp
Genomic Map