Assay Details
Gene Symbol: Rapgef5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK043108.1 | - | 3 | 2245 | 110 | |
AK087347.1 | - | 1 | 321 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
