Assay Details
Gene Symbol: Gm5486 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK135986.1 | - | 1 - 2 | 159 | 93 | |
AK139943.1 | - | 186 | 93 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK139943.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
