Assay Details
| Gene Symbol: Lnpep | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK147232.1 | - | 84 | |||
| AK155945.1 | - | 4620 | 84 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lnpep| Gene Symbol: Lnpep | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK147232.1 | - | 84 | |||
| AK155945.1 | - | 4620 | 84 | |||