Assay Details
| Gene Symbol: Capn15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK079422.1 | - | 9 | 82 | ||
| AK143711.1 | - | 12 | 82 | |||
| AK170543.1 | - | 11 | 82 | |||
| AK191640.1 | - | 1 | 231 | 82 | ||
| AK192933.1 | - | 1 | 82 | |||
| AK194114.1 | - | 1 | 234 | 82 | ||
| AK194322.1 | - | 1 | 234 | 82 | ||
| BC058094.1 | - | 10 | 6824892 | 82 | ||
More Information
Set Membership: |
|

Capn15
Genomic Map