Assay Details
| Gene Symbol: Tulp4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103181.1 | NP_001096651.1 | 13 - 14 | 4439 | 72 | |
| NM_054040.3 | NP_473381.1 | 13 - 14 | 5281 | 72 | ||
| GenBank mRNA | AF219945.1 | - | 13 - 14 | 4685 | 72 | |
| AK039439.1 | - | 1 - 2 | 1163 | 72 | ||
| AK147643.1 | - | 13 - 14 | 4379 | 72 | ||
| BC008557.1 | - | 1 - 2 | 652 | 72 | ||
| BC138024.1 | - | 13 - 14 | 4616 | 8861043 | 72 | |
| BC138025.1 | - | 13 - 14 | 4616 | 8861044 | 72 | |
More Information
Set Membership: |
|

Tulp4
Genomic Map