Assay Details
| Gene Symbol: Stk-ps2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_037690.1 | 1 | 1128 | 85 | ||
| NR_037691.1 | 1 | 1128 | 85 | |||
| GenBank mRNA | AK029722.1 | - | 1 | 1161 | 85 | |
| BC145241.1 | - | 1 | 1128 | 9053793 | 85 | |
| BC145242.1 | - | 1 | 1128 | 9053794 | 85 | |
| BC150733.1 | - | 1 | 1128 | 9087644 | 85 | |
More Information
Set Membership: |
|

Stk-ps2
Genomic Map