Assay Details
| Gene Symbol: Nutf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026532.3 | NP_080808.1 | 5 | 517 | 96 | |
| GenBank mRNA | AK002461.1 | - | 5 | 499 | 96 | |
| AK012500.1 | - | 5 | 499 | 96 | ||
| AK020512.1 | - | 5 | 516 | 96 | ||
| AK028410.1 | - | 5 | 507 | 96 | ||
| AK051049.1 | - | 5 | 499 | 96 | ||
| AK152212.1 | - | 5 | 97 | |||
| AK168026.1 | - | 5 | 499 | 96 | ||
| BC003955.1 | - | 5 | 517 | 3487899 | 96 | |
| BC083165.1 | - | 5 | 571 | 6437914 | 96 | |
| BC086773.1 | - | 5 | 484 | 6828661 | 96 | |
| Gene Symbol: Nutf2-ps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_033574.1 | 1 | 498 | 96 | ||
| GenBank mRNA | CJ235788.1 | - | 1 | 100 | 96 | |
More Information
Set Membership: |
|

Nutf2
Genomic Map