Assay Details
| Gene Symbol: Fabp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001272097.1 | NP_001259026.1 | 4 | 580 | 102 | |
| NM_001272098.1 | NP_001259027.1 | 3 | 410 | 102 | ||
| NM_010634.3 | NP_034764.1 | 4 | 583 | 102 | ||
| GenBank mRNA | AK008782.1 | - | 4 | 577 | 102 | |
| AK011551.1 | - | 4 | 579 | 102 | ||
| AK166506.1 | - | 4 | 576 | 102 | ||
| AK167389.1 | - | 4 | 578 | 102 | ||
| BC002008.1 | - | 4 | 563 | 3490535 | 102 | |
| BC053042.1 | - | 2 | 3143 | 102 | ||
| BC100543.1 | - | 4 | 558 | 30136900 | 102 | |
| W54445.1 | - | 3 | 363 | 102 | ||
| X70100.1 | - | 4 | 562 | 102 | ||
More Information
Set Membership: |
|

Fabp5
Genomic Map