Assay Details
Gene Symbol: D530039A21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AV227609.2 | - | 1 | 182 | 83 |
Gene Symbol: Slc25a37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK034948.1 | - | 2595 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
