Assay Details
| Gene Symbol: Impa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK054245.1 | - | 1524 | 105 | ||
| AK080544.1 | - | 2391 | 105 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Impa2| Gene Symbol: Impa2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK054245.1 | - | 1524 | 105 | ||
| AK080544.1 | - | 2391 | 105 | |||
Set Membership: |
|