Assay Details
| Gene Symbol: Riok3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK053740.1 | - | 89 | |||
| AK054308.1 | - | 1 | 222 | 89 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Riok3| Gene Symbol: Riok3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK053740.1 | - | 89 | |||
| AK054308.1 | - | 1 | 222 | 89 | ||
Set Membership: |
|