Assay Details
| Gene Symbol: Cycs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_007808.4 | NP_031834.1 | 3 | 611 | 75 | |
| GenBank mRNA | AK002732.1 | - | 3 | 613 | 75 | |
| AK003168.1 | - | 3 | 640 | 75 | ||
| AK088098.1 | - | 3 | 612 | 75 | ||
| AK143887.1 | - | 3 | 612 | 75 | ||
| BC034363.1 | - | 3 | 595 | 2650742 | 75 | |
| BC094054.1 | - | 3 | 600 | 2865963 | 75 | |
More Information
Set Membership: |
|

Cycs
Genomic Map