Assay Details
| Gene Symbol: Ccrn4l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | U70139.1 | - | 2 | 1630 | 91 | |
| Gene Symbol: BC094435 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK088963.1 | - | 1 | 539 | 91 | |
| BC094435.1 | - | 1 | 573 | 91 | ||
| Gene Symbol: Sgip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK090117.1 | - | 3567 | 91 | ||
| AK138403.1 | - | 624 | 91 | |||
More Information
Set Membership: |
|

Ccrn4l
Genomic Map