Assay Details
| Gene Symbol: Dusp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK153769.1 | - | 516 | 65 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dusp5| Gene Symbol: Dusp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK153769.1 | - | 516 | 65 | ||