Assay Details
| Gene Symbol: Glo1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025374.3 | NP_079650.3 | 6 | 873 | 103 | |
| GenBank mRNA | AK002386.1 | - | 6 | 801 | 103 | |
| AK003567.1 | - | 6 | 821 | 103 | ||
| AK005215.1 | - | 6 | 822 | 103 | ||
| AK031832.1 | - | 6 | 815 | 103 | ||
| AK049703.1 | - | 6 | 830 | 103 | ||
| BC024663.1 | - | 6 | 833 | 3591599 | 103 | |
More Information
Set Membership: |
|

Glo1
Genomic Map