Assay Details
| Gene Symbol: Ptpmt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025576.2 | NP_079852.1 | 1 | 294 | 95 | |
| GenBank mRNA | AK012674.1 | - | 1 | 283 | 95 | |
| AK014691.1 | - | 1 | 288 | 95 | ||
| AK151972.1 | - | 1 | 295 | 95 | ||
| AK168416.1 | - | 102 | 95 | |||
| BC026750.1 | - | 1 | 273 | 5135841 | 95 | |
More Information
Set Membership: |
|

Ptpmt1
Genomic Map