Assay Details
| Gene Symbol: Snrpd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009226.4 | NP_033252.1 | 3 | 336 | 110 | |
| GenBank mRNA | AK005517.1 | - | 3 | 326 | 110 | |
| AK007674.1 | - | 3 | 337 | 110 | ||
| AK011455.1 | - | 3 | 330 | 110 | ||
| AK013075.1 | - | 3 | 303 | 110 | ||
| AK134582.1 | - | 3 | 330 | 110 | ||
| AK135979.1 | - | 3 | 326 | 110 | ||
| BC024875.1 | - | 3 | 322 | 4009014 | 110 | |
| M58558.1 | - | 3 | 265 | 110 | ||
More Information
Set Membership: |
|

Snrpd1
Genomic Map