Assay Details
| Gene Symbol: Kcng1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081134.1 | NP_001074603.1 | 1 | 522 | 103 | |
| GenBank mRNA | BC147251.1 | - | 1 | 454 | 9056302 | 103 |
| BC147252.1 | - | 1 | 454 | 9056297 | 103 | |
| BC147715.1 | - | 1 | 454 | 9087474 | 103 | |
More Information
Set Membership: |
|

Kcng1
Genomic Map