Assay Details
| Gene Symbol: Vim | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011701.4 | NP_035831.2 | 1 | 42 | 50 | |
| GenBank mRNA | AK076001.1 | - | 1 | 44 | 50 | |
| AK131618.1 | - | 1 | 44 | 50 | ||
| AK133726.1 | - | 1 | 43 | 50 | ||
| AK151196.1 | - | 1 | 45 | 50 | ||
| AK153019.1 | - | 1 | 45 | 50 | ||
| AK159539.1 | - | 1 | 43 | 50 | ||
| AK169181.1 | - | 1 | 44 | 50 | ||
| AK170429.1 | - | 1 | 43 | 50 | ||
| BC089335.1 | - | 1 | 38 | 6415629 | 50 | |
| M26251.1 | - | 1 | 401 | 50 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK131618.1, AK133726.1, AK151196.1, AK153019.1, AK159539.1, AK169181.1, AK170429.1, BC089335.1, M26251.1, NM_011701.4 transcripts.
Set Membership: |
|

Vim
Genomic Map