Assay Details
| Gene Symbol: Zswim3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_178375.2 | NP_848462.1 | 2 | 1278 | 90 | |
| GenBank mRNA | AK016241.1 | - | 1 | 90 | ||
| BC023287.1 | - | 2 | 1278 | 3993352 | 90 | |
More Information
Set Membership: |
|

Zswim3
Genomic Map