Assay Details
| Gene Symbol: Rpl6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011290.5 | NP_035420.2 | 2 | 152 | 89 | |
| GenBank mRNA | AK002248.1 | - | 2 | 118 | 89 | |
| AK002648.1 | - | 2 | 116 | 89 | ||
| AK010918.1 | - | 1 | 111 | 89 | ||
| AK088472.1 | - | 1 | 119 | 89 | ||
| AK135175.1 | - | 2 | 121 | 89 | ||
| AK148479.1 | - | 2 | 120 | 89 | ||
| AK150540.1 | - | 2 | 124 | 89 | ||
| AK151022.1 | - | 2 | 124 | 89 | ||
| AK151344.1 | - | 2 | 124 | 89 | ||
| AK151759.1 | - | 2 | 124 | 89 | ||
| AK153208.1 | - | 2 | 153 | 89 | ||
| AK153507.1 | - | 2 | 124 | 89 | ||
| AK167439.1 | - | 2 | 124 | 89 | ||
| BC062880.1 | - | 1 | 106 | 5692572 | 89 | |
| BC106121.1 | - | 1 | 105 | 3979296 | 89 | |
| X81987.1 | - | 1 | 100 | 89 | ||
More Information
Set Membership: |
|

Rpl6
Genomic Map