Assay Details
| Gene Symbol: Gpc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016696.4 | NP_057905.1 | 1 | 138 | 87 | |
| GenBank mRNA | AF185613.1 | - | 1 | 141 | 87 | |
| AK154899.1 | - | 1 | 181 | 87 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AF185613.1, AK154899.1, NM_016696.4 transcripts.
Set Membership: |
|

Gpc1
Genomic Map