Assay Details
| Gene Symbol: Gal3st3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001024717.2 | NP_001019888.1 | 3 | 1063 | 62 | |
| GenBank mRNA | BC066996.1 | - | 3 | 1053 | 5707267 | 62 |
More Information
Set Membership: |
|

Gal3st3
Genomic Map