Assay Details
| Gene Symbol: Bri3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001163709.1 | NP_001157181.1 | 3 | 462 | 100 | |
| NM_018772.4 | NP_061242.3 | 3 | 578 | 100 | ||
| GenBank mRNA | AF106967.1 | - | 3 | 497 | 100 | |
| AK080467.1 | - | 3 | 568 | 100 | ||
| BC005677.1 | - | 3 | 556 | 100 | ||
| BC021149.1 | - | 3 | 524 | 100 | ||
| BC095955.1 | - | 3 | 533 | 5328625 | 100 | |
| BE991935.1 | - | 1 | 100 | |||
| X61454.1 | - | 2 | 226 | 100 | ||
More Information
Set Membership: |
|

Bri3
Genomic Map