Assay Details
| Gene Symbol: Phyhip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145981.3 | NP_666093.1 | 5 | 2036 | 79 | |
| GenBank mRNA | AK049900.1 | - | 5 | 2037 | 79 | |
| BC030494.1 | - | 5 | 2033 | 79 | ||
More Information
Set Membership: |
|

Phyhip
Genomic Map