Assay Details
| Gene Symbol: Cst3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_009976.3 | NP_034106.2 | 3 | 524 | 92 | |
| GenBank mRNA | AK002438.1 | - | 3 | 525 | 92 | |
| AK013676.1 | - | 3 | 493 | 92 | ||
| AK014368.1 | - | 3 | 544 | 92 | ||
| AK131728.1 | - | 3 | 542 | 92 | ||
| AK146333.1 | - | 3 | 553 | 92 | ||
| AK151160.1 | - | 3 | 489 | 92 | ||
| AK161856.1 | - | 3 | 487 | 92 | ||
| AK166430.1 | - | 3 | 516 | 92 | ||
| AK212642.1 | - | 3 | 241 | 92 | ||
| BC002072.1 | - | 3 | 470 | 3593373 | 92 | |
| M59470.1 | - | 3 | 457 | 92 | ||
More Information
Set Membership: |
|

Cst3
Genomic Map