Assay Details
| Gene Symbol: Ppp4r1l-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK030794.1 | - | 1 | 1601 | 66 | |
| AK046376.1 | - | 1 | 1276 | 66 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppp4r1l-ps| Gene Symbol: Ppp4r1l-ps | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK030794.1 | - | 1 | 1601 | 66 | |
| AK046376.1 | - | 1 | 1276 | 66 | ||
Set Membership: |
|