Assay Details
| Gene Symbol: Basp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027395.2 | NP_081671.1 | 2 | 1548 | 129 | |
| GenBank mRNA | AK011545.1 | - | 2 | 1511 | 129 | |
| AK046868.1 | - | 2 | 1524 | 129 | ||
| AK159486.1 | - | 3 | 939 | 129 | ||
More Information
Set Membership: |
|

Basp1
Genomic Map