Assay Details
Gene Symbol: Rgs5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK028765.1 | - | 2044 | 82 | ||
AK030300.1 | - | 2043 | 82 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rgs5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK028765.1 | - | 2044 | 82 | ||
AK030300.1 | - | 2043 | 82 |