Assay Details
| Gene Symbol: Rae1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175112.5 | NP_780321.1 | 12 | 84 | ||
| GenBank mRNA | AK014325.1 | - | 12 | 84 | ||
| AK017746.1 | - | 12 | 84 | |||
| AK079383.1 | - | 12 | 84 | |||
| AK146058.1 | - | 12 | 84 | |||
| AK156681.1 | - | 12 | 84 | |||
| BC059051.1 | - | 12 | 6818771 | 84 | ||
| BC060072.1 | - | 10 | 5718934 | 84 | ||
| CO041965.1 | - | 1 | 198 | 84 | ||
More Information
Set Membership: |
|

Rae1
Genomic Map