Assay Details
| Gene Symbol: Pbxip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_146131.2 | NP_666243.1 | 11 | 3069 | 89 | |
| GenBank mRNA | AK047898.1 | - | 2 | 914 | 89 | |
| AK081345.1 | - | 2 | 914 | 89 | ||
| AK143270.1 | - | 1 | 702 | 89 | ||
More Information
Set Membership: |
|

Pbxip1
Genomic Map