Assay Details
| Gene Symbol: Papss1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289477.1 | NP_001276406.1 | 13 | 164 | ||
| NM_001289478.1 | NP_001276407.1 | 13 | 164 | |||
| NM_001289479.1 | NP_001276408.1 | 14 | 164 | |||
| NM_011863.2 | NP_035993.1 | 12 | 164 | |||
| GenBank mRNA | AK146507.1 | - | 12 | 164 | ||
| AK153296.1 | - | 12 | 164 | |||
| AK160875.1 | - | 14 | 164 | |||
| AK168982.1 | - | 12 | 164 | |||
| AK169117.1 | - | 13 | 164 | |||
| AK169124.1 | - | 12 | 164 | |||
| AK169204.1 | - | 13 | 164 | |||
| BC066055.1 | - | 13 | 6833756 | 164 | ||
| CO043332.1 | - | 1 | 103 | 165 | ||
| U34883.1 | - | 12 | 164 | |||
More Information
Set Membership: |
|

Papss1
Genomic Map