Assay Details
| Gene Symbol: Topors | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134097.3 | NP_598858.2 | 3 | 3709 | 125 | |
| GenBank mRNA | AB072395.1 | - | 3 | 3667 | 125 | |
| AB104865.1 | - | 3 | 3695 | 125 | ||
| BC040797.1 | - | 3 | 3696 | 3492178 | 125 | |
More Information
Set Membership: |
|

Topors
Genomic Map