Assay Details
| Gene Symbol: Ppp2r5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144880.4 | NP_659129.2 | 3 | 1041 | 71 | |
| GenBank mRNA | AK145397.1 | - | 4 | 427 | 71 | |
| BC059026.1 | - | 3 | 1041 | 6810754 | 71 | |
More Information
Set Membership: |
|

Ppp2r5a
Genomic Map