Assay Details
| Gene Symbol: Card10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_130859.2 | NP_570929.2 | 21 | 3102 | 83 | |
| GenBank mRNA | AF363456.1 | - | 20 | 2951 | 83 | |
| AK079433.1 | - | 5 | 538 | 83 | ||
| BC060203.1 | - | 21 | 3102 | 6855024 | 83 | |
More Information
Set Membership: |
|

Card10
Genomic Map