Assay Details
| Gene Symbol: Tmeff1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021436.2 | NP_067411.1 | 10 | 2031 | 81 | |
| GenBank mRNA | AK079633.1 | - | 8 | 1380 | 81 | |
| BC023714.1 | - | 5 | 1193 | 81 | ||
| BC057598.1 | - | 10 | 2031 | 6401547 | 81 | |
More Information
Set Membership: |
|

Tmeff1
Genomic Map