Assay Details
| Gene Symbol: Ugcg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011673.3 | NP_035803.1 | 9 | 2651 | 92 | |
| GenBank mRNA | BC050828.1 | - | 9 | 2646 | 6512920 | 92 |
More Information
Set Membership: |
|

Ugcg
Genomic Map