Assay Details
| Gene Symbol: Ccnl2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207678.1 | NP_997561.1 | 11 | 2046 | 88 | |
| GenBank mRNA | AB041605.1 | - | 11 | 2023 | 88 | |
| AK008585.1 | - | 6 | 1040 | 88 | ||
| AK048244.1 | - | 11 | 3826 | 88 | ||
| BC023747.1 | - | 11 | 3814 | 88 | ||
| BC083055.1 | - | 4 | 1033 | 88 | ||
| BC132295.1 | - | 11 | 2038 | 40130572 | 88 | |
| U37351.1 | - | 4 | 1052 | 88 | ||
More Information
Set Membership: |
|

Ccnl2
Genomic Map