Assay Details
| Gene Symbol: Paxip1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018878.3 | NP_061366.2 | 21 | 3559 | 77 | |
| GenBank mRNA | AF104261.1 | - | 21 | 3559 | 77 | |
| AK144606.1 | - | 21 | 3421 | 77 | ||
| AK148141.1 | - | 21 | 3432 | 77 | ||
| BC066014.1 | - | 21 | 3348 | 5704566 | 77 | |
More Information
Set Membership: |
|

Paxip1
Genomic Map