Assay Details
| Gene Symbol: Slain2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113423.2 | NP_001106894.1 | 7 | 1681 | 60 | |
| NM_153567.3 | NP_705795.2 | 8 | 1759 | 60 | ||
| GenBank mRNA | AK129365.1 | - | 7 | 1670 | 60 | |
| AK146432.1 | - | 7 | 1680 | 60 | ||
| BC019454.1 | - | 1 | 60 | |||
| BC038019.1 | - | 6 | 964 | 3597423 | 60 | |
| BC076573.1 | - | 7 | 1614 | 60 | ||
| CK619668.1 | - | 4 | 466 | 60 | ||
More Information
Set Membership: |
|

Slain2
Genomic Map