Assay Details
| Gene Symbol: Cabs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027631.2 | NP_081907.1 | 1 | 616 | 93 | |
| GenBank mRNA | AK016441.1 | - | 1 | 623 | 93 | |
| AK076636.1 | - | 1 | 617 | 93 | ||
| BC049750.1 | - | 1 | 616 | 6773045 | 93 | |
More Information
Set Membership: |
|

Cabs1
Genomic Map