Assay Details
| Gene Symbol: Mrps17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025450.4 | NP_079726.1 | 4 | 901 | 115 | |
| GenBank mRNA | AK003008.1 | - | 4 | 718 | 115 | |
| AK007356.1 | - | 4 | 902 | 115 | ||
| AK009568.1 | - | 4 | 899 | 115 | ||
| AK134065.1 | - | 3 | 1690 | 115 | ||
| BC003890.1 | - | 3 | 838 | 3592661 | 115 | |
More Information
Set Membership: |
|

Mrps17
Genomic Map