Assay Details
| Gene Symbol: Gsdmd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026960.4 | NP_081236.1 | 1 | 40 | 115 | |
| GenBank mRNA | AK007710.1 | - | 1 | 41 | 115 | |
| AK088747.1 | - | 1 | 52 | 115 | ||
| AK165858.1 | - | 1 | 41 | 115 | ||
| AK171294.1 | - | 1 | 42 | 115 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK007710.1, AK165858.1, AK171294.1, NM_026960.4 transcripts.
Set Membership: |
|

Gsdmd
Genomic Map